Få mere at vide
1,2-Dihydroxybenzene-d6, CDN
Artikelnummer. 30313688
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30313688 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313688 Leverandør CDN Leverandørnr. CDND52901G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Catechol-d6
CAS 202656-22-2
Molekylær formel C6 2H6 O2
Mængde 1 g
Synonym 1,2-Benzene-3,4,5,6-d4-diol-d2, Pyrogallol-d6, 1,2-Benzenediol-1,2,3,4,5,6-D6, Catechol-1,2,3,4,5,6-D6, Catechol D6, Benzene-1,2-diol D6
InChI formel InChI=1 S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8 H/i1D,2 D,3 D,4 D/hD2
SMIL [2 H]Oc1c([2 H])c([2 H])c([2 H])c([2 H])c1O[2 H]
IUPAC navn 1,2,3,4-tetradeuterio-5,6-dideuteriooxybenzene
Molekylvægt (g/mol) 116.15
Formel vægt 116.0744 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity