Få mere at vide
1,2-Dihydro Fluticasone Propionate, TRC
Artikelnummer. 30011135
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011135 100 mg
30006814 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011135 Leverandør Toronto Research Chemicals Leverandørnr. TRCD449635100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 105613-90-9
Molekylær formel C25 H33 F3 O5 S
Molekylvægt (g/mol) 502.59
Synonym Androst-4-ene-17-carbothioic acid, 6,9-difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)-, S-(fluoromethyl) ester, (6α,11β,16α,17α)-, Fluticasone Propionate Imp. E (EP), 1,2-dihydro fluticasone propionate, GR 51610X, unidentified impurity with RRT 1.23, disulphide dipropionate dithioketone (GR 247095X), dithio (GR 269949X)
IUPAC navn [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-17-(fluoromethylsulfanylcarbonyl)-11-hydroxy-10,13,16-trimethyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] propanoate
SMIL CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SCF
Tekniske data
Kemisk navn eller materiale 1,2-Dihydro Fluticasone Propionate
Mængde 100 mg
InChI formel InChI=1S/C25H33F3O5S/c1-5-20(31)33-25(21(32)34-12-26)13(2)8-15-16-10-18(27)17-9-14(29)6-7-22(17,3)24(16,28)19(30)11-23(15,25)4/h9,13,15-16,18-19,30H,5-8,10-12H2,1-4H3/t13-,15+,16+,18+,19+,22+,23+,24+,25+/m1/s1
Formel vægt 502.2001