Learn More
1,2-Difluorobenzene, 98+%, Thermo Scientific Chemicals
Catalog Number p-7080728
Click to view available options
Quantity:
10 g
50 g
250 g
367-11-3
C6H4F2
114.10
MFCD00000284
GOYDNIKZWGIXJT-UHFFFAOYSA-N
o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene
9706
CHEBI:38583
1,2-difluorobenzene
FC1=CC=CC=C1F
367-11-3
C6H4F2
114.10
MFCD00000284
GOYDNIKZWGIXJT-UHFFFAOYSA-N
o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene
9706
CHEBI:38583
1,2-difluorobenzene
FC1=CC=CC=C1F
1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY
Melting Point -34°C
Density 1.171
Boiling Point 92°C to 94°C
Flash Point 1°C (33°F)
Odor Characteristic
Refractive Index 1.444
Quantity 10 g
UN Number UN1993
Beilstein 1905113
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 114.09
Percent Purity ≥98%
Chemical Name or Material 1,2-Difluorobenzene