Få mere at vide
(±)-1,2-Dichloropropane-d6, CDN
Artikelnummer. 30327666
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327666 0.25 g -
30309345 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327666 Leverandør CDN Leverandørnr. CDND23630.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Dichloropropane-d6
CAS 93952-08-0
Molekylær formel C3 D6 Cl2
Mængde 0.25 g
InChI formel InChI=1 S/C3H6Cl2/c1-3(5)2-4/h3H,2H2,1H3/i1D3,2D2,3 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(Cl)C([2 H])([2 H])Cl
IUPAC navn 1,2-dichloro-1,1,2,3,3,3-hexadeuterio-propane
Molekylvægt (g/mol) 119.02
Formel vægt 118.0223 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity