missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromotetrachloroethane, 97%, Thermo Scientific Chemicals

Catalog Number p-7024991
179.35 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
630-25-1
C2Br2Cl4
325.63
MFCD00000780
WJUKOGPNGRUXMG-UHFFFAOYSA-N
1,2-dibromotetrachloroethane, dbtce, sym-dibromotetrachloroethane, ethane, 1,2-dibromo-1,1,2,2-tetrachloro, unii-0sgg217efr, 0sgg217efr, 1,2-dibrom-1,1,2,2-tetrachlorethan, brccl2ccl2br, 1,2-dibromotetrachloroethan, 1,2-dibrom-tetrachlorethane
69426
1,2-dibromo-1,1,2,2-tetrachloroethane
C(C(Cl)(Cl)Br)(Cl)(Cl)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
630-25-1
C2Br2Cl4
325.63
MFCD00000780
WJUKOGPNGRUXMG-UHFFFAOYSA-N
1,2-dibromotetrachloroethane, dbtce, sym-dibromotetrachloroethane, ethane, 1,2-dibromo-1,1,2,2-tetrachloro, unii-0sgg217efr, 0sgg217efr, 1,2-dibrom-1,1,2,2-tetrachlorethan, brccl2ccl2br, 1,2-dibromotetrachloroethan, 1,2-dibrom-tetrachlorethane
69426
1,2-dibromo-1,1,2,2-tetrachloroethane
C(C(Cl)(Cl)Br)(Cl)(Cl)Br

1,2-dibromotetrachloroethane is used as a fire-proofing agent. It is actively involved in the conversion of sulfones to alkenes by Ramberg-Backlund rearrangement. It is an effective replacement for ozone depleting solvents like dibromodifluoromethane and tetrabromodifluoroethane. It is also utilized as a brominating reagent. In analytical chemistry, it is used in quantitative determination of perhalogenated compounds by high-performance liquid chromatography (HPLC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-dibromotetrachloroethane is used as a fire-proofing agent. It is actively involved in the conversion of sulfones to alkenes by Ramberg-Backlund rearrangement. It is an effective replacement for ozone depleting solvents like dibromodifluoromethane and tetrabromodifluoroethane. It is also utilized as a brominating reagent. In analytical chemistry, it is used in quantitative determination of perhalogenated compounds by high-performance liquid chromatography (HPLC).

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼220°C (decomposition)
Density 2.71
Quantity 25 g
Beilstein 1699471
Solubility Information Insoluble in water.
Formula Weight 325.65
Percent Purity 98%
Chemical Name or Material 1,2-Dibromotetrachloroethane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'