Få mere at vide
1,2-Dibromobenzene-d4, CDN
Artikelnummer. 30311839
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311839 0.5 g
30323435 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311839 Leverandør CDN Leverandørnr. CDND78390.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Dibromobenzene-D4
CAS 1643577-34-7
Molekylær formel C6 D4 Br2
Mængde 0.5 g
Synonym Benzene-1,2,3,4-d4, 5,6-dibromo- (ACI), 5,6-Dibromobenzene-1,2,3,4-d4 (ACI), 1,2-Dibromobenzene-d4, 1,2-Dibromobenzene-D4
InChI formel InChI=1 S/C6H4Br2/c7-5-3-1-2-4-6(5)8/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c([2 H])c(Br)c(Br)c1[2 H]
IUPAC navn 1,2-dibromo-3,4,5,6-tetradeuteriobenzene
Molekylvægt (g/mol) 239.9286
Formel vægt 237.8931 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity