Få mere at vide
1,2-Cyclohexanedicarboxylic Acid Mono 4-Methyloctyl Ester, TRC
Artikelnummer. 30007302
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007302 25 mg
30007271 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30007302 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98730525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1889286-78-5
Molekylær formel C17H30O4
Molekylvægt (g/mol) 298.42
Synonym 2-(((4-Methyloctyl)oxy)carbonyl)cyclohexanecarboxylic Acid, MINCH
IUPAC navn 2-(((4-methyloctyl)oxy)carbonyl)cyclohexane-1-carboxylic acid
SMIL O=C(O)C1C(C(OCCCC(C)CCCC)=O)CCCC1
Tekniske data
Kemisk navn eller materiale 1,2-Cyclohexanedicarboxylic Acid Mono 4-Methyloctyl Ester
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C17H30O4/c1-3-4-8-13(2)9-7-12-21-17(20)15-11-6-5-10-14(15)16(18)19/h13-15H,3-12H2,1-2H3,(H,18,19)
Formel vægt 298.21
Procent renhed >95%