missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Chlorophenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62129.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15408708

  • 2403.83 DKK / 1g


Estimated Shipment date
from Supplier 01-08-2018
Add to basket

Specifications

Chemical Identifiers

1-(2-Chlorophenyl)-3-(4-fluorophenyl)urea
MFCD00018186

Specifications

1-(2-Chlorophenyl)-3-(4-fluorophenyl)urea
1g
97%
264.68
Solid
MFCD00018186
Videos
SDS