Få mere at vide
1-(2-Chloroethyl)-3-cyclohexylurea, Mikromol™
Artikelnummer. 30050689
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050689 25 mg
17972886 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050689 Leverandør Mikromol Leverandørnr. MM0923.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(2-Chloroethyl)-3-cyclohexylurea
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(2-Chloroethyl)-3-cyclohexylurea
CAS 13908-11-7
Analyt- eller komponentnavne 1-(2-Chloroethyl)-3-cyclohexylurea
Forsendelsestilstand Room Temperature
Molekylær formel C9 H17 Cl N2 O
Certificeringer/compliance ISO 9001
Synonym Lomustine Imp. B (EP), Lomustine USP Related Compound B, Lomustine USP RC B, 1-(2-Chloroethyl)-3-cyclohexylurea, Lomustine Related Compound B, Lomustine Impurity B
InChI formel InChI=1S/C9H17ClN2O/c10-6-7-11-9(13)12-8-4-2-1-3-5-8/h8H,1-7H2,(H2,11,12,13)
SMIL ClCCNC(=O)NC1CCCCC1
IUPAC navn 1-(2-chloroethyl)-3-cyclohexylurea
Molekylvægt (g/mol) 204.7
Mængde 25 mg
Formel vægt 204.1029 g/mol