missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)piperidine, 97%

Brand:  Alfa Aesar™ H63283.03

Additional Details : CAS Number : 3970-42-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489898

  • 669.72 DKK / 1g


Estimated Shipment
20-10-2020
Add to basket

Specifications

Chemical Identifiers

3970-42-1
240.687
1-2-chloro-6-nitrophenyl piperidine, 1-2-chloro-6-nitro-phenyl-piperidine, 6-chloro-2-nitrophenyl piperidine
1-(2-chloro-6-nitrophenyl)piperidine
C11H13ClN2O2
IBRSUFPYKBRXMU-UHFFFAOYSA-N
4778967
C1CCN(CC1)C2=C(C=CC=C2Cl)[N+](=O)[O-]

Specifications

1-(2-Chloro-6-nitrophenyl)piperidine
3970-42-1
1317515
IBRSUFPYKBRXMU-UHFFFAOYSA-N
1-(2-chloro-6-nitrophenyl)piperidine
4778967
Solid
1g
C11H13ClN2O2
1-2-chloro-6-nitrophenyl piperidine, 1-2-chloro-6-nitro-phenyl-piperidine, 6-chloro-2-nitrophenyl piperidine
C1CCN(CC1)C2=C(C=CC=C2Cl)[N+](=O)[O-]
240.687
240.69
97%
Videos
SDS Documents