Learn More
Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)piperidine, 97%
Brand: Alfa Aesar™ H63283.06
Additional Details : CAS Number : 3970-42-1 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
3970-42-1 | |
240.687 | |
1-2-chloro-6-nitrophenyl piperidine, 1-2-chloro-6-nitro-phenyl-piperidine, 6-chloro-2-nitrophenyl piperidine | |
1-(2-chloro-6-nitrophenyl)piperidine |
C11H13ClN2O2 | |
IBRSUFPYKBRXMU-UHFFFAOYSA-N | |
4778967 | |
C1CCN(CC1)C2=C(C=CC=C2Cl)[N+](=O)[O-] |
Specifications
1-(2-Chloro-6-nitrophenyl)piperidine | |
3970-42-1 | |
1317515 | |
IBRSUFPYKBRXMU-UHFFFAOYSA-N | |
1-(2-chloro-6-nitrophenyl)piperidine | |
4778967 | |
Solid |
5g | |
C11H13ClN2O2 | |
1-2-chloro-6-nitrophenyl piperidine, 1-2-chloro-6-nitro-phenyl-piperidine, 6-chloro-2-nitrophenyl piperidine | |
C1CCN(CC1)C2=C(C=CC=C2Cl)[N+](=O)[O-] | |
240.687 | |
240.69 | |
97% |