Få mere at vide
1-(2-Chloro-4-(4-chlorophenoxy)phenyl)-2-(1H-1,2,4-triazol-1-yl)ethan-1-ol, TRC
Artikelnummer. 30357082
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30357082 10 mg
30350216 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30357082 Leverandør Toronto Research Chemicals Leverandørnr. TRCC42925310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 117018-19-6
Molekylær formel C16 H13 Cl2 N3 O2
Molekylvægt (g/mol) 350.2
Synonym α-[2-Chloro-4-(4-chlorophenoxy)phenyl]-1H-1,2,4-triazole-1-ethanol, CGA 205375
IUPAC navn 1-[2-chloro-4-(4-chlorophenoxy)phenyl]-2-(1,2,4-triazol-1-yl)ethanol
SMIL OC(Cn1cncn1)c2ccc(Oc3ccc(Cl)cc3)cc2Cl
Tekniske data
Kemisk navn eller materiale Difenoconazole-alcohol
Mængde 10 mg
InChI formel InChI=1S/C16H13Cl2N3O2/c17-11-1-3-12(4-2-11)23-13-5-6-14(15(18)7-13)16(22)8-21-10-19-9-20-21/h1-7,9-10,16,22H,8H2
Formel vægt 349.0385