Learn More
Alfa Aesar™ 1-(2-Bromophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%
Brand:  Alfa Aesar™ H65433.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15469636
  • 1882.87 DKK / 1g
Estimated Shipment date
from Supplier 12-03-2018
Add to basket
Specifications
Chemical Identifiers
1-(2-Bromophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
393.59
MFCD27975762
Specifications
1-(2-Bromophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
Solid
393.59
1g
97%
MFCD27975762