missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(trimethylsilyloxy)ethane, 98%, Thermo Scientific Chemicals

Catalog Number p-3756938
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7381-30-8
C8H22O2Si2
206.43
MFCD00009640
JGWFUSVYECJQDT-UHFFFAOYSA-N
1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50
81858
trimethyl(2-trimethylsilyloxyethoxy)silane
C[Si](C)(C)OCCO[Si](C)(C)C
This item is not returnable. View return policy
7381-30-8
C8H22O2Si2
206.43
MFCD00009640
JGWFUSVYECJQDT-UHFFFAOYSA-N
1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50
81858
trimethyl(2-trimethylsilyloxyethoxy)silane
C[Si](C)(C)OCCO[Si](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 165°C to 166°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3SiOCH2CH2OSi(CH3)3
Refractive Index 1.4020 to 1.404
Quantity 10 g
Specific Gravity 0.84
Formula Weight 206.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Bis(trimethylsilyloxy)ethane
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Bis(trimethylsilyloxy)ethane
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'