missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dimethylsilyl)benzene, 98%, Thermo Scientific Chemicals

Catalog Number p-7059004
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
17985-72-7
C10H16Si2
192.408
MFCD00142462
MUUXBTFQEXVEEI-UHFFFAOYSA-N
1,2-bis dimethylsilyl benzene, 1,2-phenylenebis dimethylsilane, 2-dimethyl, acmc-209efy, bis dimethylsilyl benzene, amtsi084, o-phenylenebis dimethylsilane, 2-dimethylsilylphenyl-dimethylsilane, 5,6-bis dimethylsilanediyl-1,3-cyclohexadiene
6364986
[2-(dimethyl-$l^{3}-silanyl)phenyl]-dimethylsilicon
C[Si](C)C1=CC=CC=C1[Si](C)C
This item is not returnable. View return policy
17985-72-7
C10H16Si2
192.408
MFCD00142462
MUUXBTFQEXVEEI-UHFFFAOYSA-N
1,2-bis dimethylsilyl benzene, 1,2-phenylenebis dimethylsilane, 2-dimethyl, acmc-209efy, bis dimethylsilyl benzene, amtsi084, o-phenylenebis dimethylsilane, 2-dimethylsilylphenyl-dimethylsilane, 5,6-bis dimethylsilanediyl-1,3-cyclohexadiene
6364986
[2-(dimethyl-$l^{3}-silanyl)phenyl]-dimethylsilicon
C[Si](C)C1=CC=CC=C1[Si](C)C

1,2-Bis(dimethylsilyl)benzene is a protecting group-reagent for amines and amino acids forming benzostabase derivatives. It is an important raw material and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.898 g/mL
Boiling Point 101°C (13 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.514
Quantity 0.5 g
Solubility Information Insoluble in water.
Formula Weight 194.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,2-Bis (dimethylsilyl)benzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'