Få mere at vide
(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 97%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 047087.MC
| Mængde | 100mg |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| 136705-77-6 | |
| 722.63 | |
| XGPXBCKGQLCHDW-KYOOHHHUSA-M | |
| 5702639 | |
| CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC.C1CC=CCCC=C1.C(F)(F)(F)S(=O)(=O)[O-].[Rh] |
| C31H48F3O3P2RhS- | |
| MFCD00269860 | |
| --1,2-bis 2r,5r-2,5-diethylphospholano benzene cyclooctadiene rhodium i trifluoromethanesulfonate, --1,2-bis 2r,5r-2,5-diethylphospholono benzene cycloactadiene rhodium i trifluoromethanesulfonate, 1z,5z-cycloocta-1,5-diene; 2r,5r-1-2-2r,5r-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane; rhodium; trifluoromethanesulfonate | |
| (1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;trifluoromethanesulfonate |
Tekniske data
| [(R,R)Ethyl-DuPhosRh(COD)]CF3SO3 | |
| C31H48F3O3P2RhS- | |
| 100mg | |
| --1,2-bis 2r,5r-2,5-diethylphospholano benzene cyclooctadiene rhodium i trifluoromethanesulfonate, --1,2-bis 2r,5r-2,5-diethylphospholono benzene cycloactadiene rhodium i trifluoromethanesulfonate, 1z,5z-cycloocta-1,5-diene; 2r,5r-1-2-2r,5r-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane; rhodium; trifluoromethanesulfonate | |
| CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC.C1CC=CCCC=C1.C(F)(F)(F)S(=O)(=O)[O-].[Rh] | |
| 722.63 | |
| 722.62 |
| 136705-77-6 | |
| MFCD00269860 | |
| Air Sensitive | |
| XGPXBCKGQLCHDW-KYOOHHHUSA-M | |
| (1Z,5Z)-cycloocta-1,5-diene;(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane;rhodium;trifluoromethanesulfonate | |
| 5702639 | |
| 97% |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.