missing translation for 'onlineSavingsMsg'
Få mere at vide

[1-(2-Benzyloxymethyl-pyrrolidine-1-carbonyl)-2-methyl-propyl]-carbamic Acid tert-Butyl Ester, TRC

Artikelnummer. 30347655
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30347655 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30347655 Leverandør Toronto Research Chemicals Leverandørnr. TRCB286700500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-(2-Benzyloxymethyl-pyrrolidine-1-carbonyl)-2-methyl-propy
CAS 1217630-30-2
Molekylær formel C22 H34 N2 O4
Mængde 500 mg
InChI formel InChI=1S/C22H34N2O4/c1-16(2)19(23-21(26)28-22(3,4)5)20(25)24-13-9-12-18(24)15-27-14-17-10-7-6-8-11-17/h6-8,10-11,16,18-19H,9,12-15H2,1-5H3,(H,23,26)
SMIL CC(C)C(NC(=O)OC(C)(C)C)C(=O)N1CCCC1COCc2ccccc2
IUPAC navn tert-butyl N-[3-methyl-1-oxo-1-[2-(phenylmethoxymethyl)pyrrolidin-1-yl]butan-2-yl]carbamate
Molekylvægt (g/mol) 390.52
Formel vægt 390.2519
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.