Få mere at vide
1,2-Benzenedicarboxylic Acid-d4 1-(4-Hydroxybutyl) Ester, TRC
Artikelnummer. 30340135
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30340135 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340135 Leverandør Toronto Research Chemicals Leverandørnr. TRCB18545250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Benzenedicarboxylic Acid-d4 1-(4-Hydroxybutyl) Ester
CAS 1346600-32-5
Molekylær formel C12H10D4O5
Mængde 50 mg
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic Acid Mono(4-hydroxybutyl) Ester
InChI formel InChI=1S/C12H14O5/c13-7-3-4-8-17-12(16)10-6-2-1-5-9(10)11(14)15/h1-2,5-6,13H,3-4,7-8H2,(H,14,15)/i1D,2D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(C(=O)OCCCCO)c(C(=O)O)c1[2H]
IUPAC navn 2,3,4,5-tetradeuterio-6-(4-hydroxybutoxycarbonyl)benzoic acid
Molekylvægt (g/mol) 242.26
Formel vægt 242.1092