Få mere at vide
1,2-Benzenedicarboxylic Acid 1-(7-Methyloctyl) Ester-d4, TRC
Artikelnummer. 30344629
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30344629 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30344629 Leverandør Toronto Research Chemicals Leverandørnr. TRCB18550250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Benzenedicarboxylic Acid 1-(7-Methyloctyl) Ester-d4
CAS 1332965-96-4
Renhedsgrad noter HPLC
Molekylær formel C17 2H4 H20 O4
Mængde 50 mg
Synonym 1,2-Benzenedicarboxylic Acid Mono(7-methyloctyl) Ester-d4, 7-Methyl-1-Octanol Phthalate-d4
InChI formel InChI=1S/C17H24O4/c1-13(2)9-5-3-4-8-12-21-17(20)15-11-7-6-10-14(15)16(18)19/h6-7,10-11,13H,3-5,8-9,12H2,1-2H3,(H,18,19)/i6D,7D,10D,11D
SMIL [2H]c1c([2H])c([2H])c(C(=O)OCCCCCCC(C)C)c(C(=O)O)c1[2H]
IUPAC navn 2,3,4,5-tetradeuterio-6-(7-methyloctoxycarbonyl)benzoic acid
Molekylvægt (g/mol) 296.39
Formel vægt 296.1926
Procent renhed >95