missing translation for 'onlineSavingsMsg'
Learn More

1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride, 97%, Thermo Scientific™

Catalog Number p-4258959
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
866926-59-2
C24H33N2·Cl
385
RHFFYNDQCMNENO-UHFFFAOYSA-M
1-2,6-diisopropylphenyl-3-2,4,6-trimethylphenyl-imidazolinium chloride, 1-2,6-di-i-propylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride, 1-2,6-diisopropropylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride, 1-2,6-diisopropylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride, 1-2,6-diisopropylphenyl-3-mesityl-4,5-dihydro-1h-imidazol-3-ium chloride, 1-2,6-diisopropylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydro-1??-imidazol-1-ylium chloride, 3-2,6-bis 1-methylethyl phenyl-4,5-dihydro-1-2,4,6-trimethylphenyl-1h-imidazolium chloride
11617932
1-[2,6-di(propan-2-yl)phenyl]-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-3-ium;chloride
CC1=CC(=C(C(=C1)C)[N+]2=CN(CC2)C3=C(C=CC=C3C(C)C)C(C)C)C.[Cl-]
This item is not returnable. View return policy
866926-59-2
C24H33N2·Cl
385
RHFFYNDQCMNENO-UHFFFAOYSA-M
1-2,6-diisopropylphenyl-3-2,4,6-trimethylphenyl-imidazolinium chloride, 1-2,6-di-i-propylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride, 1-2,6-diisopropropylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride, 1-2,6-diisopropylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride, 1-2,6-diisopropylphenyl-3-mesityl-4,5-dihydro-1h-imidazol-3-ium chloride, 1-2,6-diisopropylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydro-1??-imidazol-1-ylium chloride, 3-2,6-bis 1-methylethyl phenyl-4,5-dihydro-1-2,4,6-trimethylphenyl-1h-imidazolium chloride
11617932
1-[2,6-di(propan-2-yl)phenyl]-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-3-ium;chloride
CC1=CC(=C(C(=C1)C)[N+]2=CN(CC2)C3=C(C=CC=C3C(C)C)C(C)C)C.[Cl-]
CAS Min % 96.0
CAS Max % 100.0
Melting Point 318°C to 319°C
Color Violet to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 100 mg
Formula Weight 385
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'