Få mere at vide
1-(2,6-Difluorobenzyl)-1H-1,2,3-triazole-4-carboxylic Acid Methyl Ester-d2, TRC
Artikelnummer. 30377259
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30377259 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377259 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44568725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(2,6-Difluorobenzyl)-1H-1,2,3-triazole-4-carboxylic Acid Methyl Ester-d2
CAS 1346597-78-1
Molekylær formel C11H7D2F2N3O2
Mængde 25 mg
Synonym 1-[(2,6-Difluorophenyl)methyl]-1H-1,2,3-triazole-4-carboxylic Acid Methyl Ester-d2
InChI formel InChI=1S/C11H9F2N3O2/c1-18-11(17)10-6-16(15-14-10)5-7-8(12)3-2-4-9(7)13/h2-4,6H,5H2,1H3/i5D2
SMIL [2H]C([2H])(c1c(F)cccc1F)n2cc(nn2)C(=O)OC
IUPAC navn methyl 1-[dideuterio-(2,6-difluorophenyl)methyl]triazole-4-carboxylate
Molekylvægt (g/mol) 255.22
Formel vægt 255.0788