Få mere at vide
1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one, Mikromol™
Artikelnummer. 17991956
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991956 25 mg
17944226 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17991956 Leverandør Mikromol Leverandørnr. MM0006.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one
CAS 15362-40-0
Analyt- eller komponentnavne 1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one
Forsendelsestilstand Room Temperature
Molekylær formel C14 H9 Cl2 N O
Certificeringer/compliance ISO 17025
Synonym 1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one, Aceclofenac Imp. I (EP), Diclofenac Sodium Imp. A (EP), Diclofenac Related Compound A (USP), Diclofenac Related Compound A, Aceclofenac Impurity I, Diclofenac Sodium Impurity A, Diclofenac Impurity A
InChI formel InChI=1S/C14H9Cl2NO/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(17)18/h1-7H,8H2
SMIL Clc1cccc(Cl)c1N2C(=O)Cc3ccccc23
IUPAC navn 1-(2,6-dichlorophenyl)-3H-indol-2-one
Molekylvægt (g/mol) 278.13
Mængde 25 mg
Formel vægt 277.0061 g/mol