missing translation for 'onlineSavingsMsg'
Learn More

1,2,5,6-Tetramethylnaphthalene, TRC

Product Code. 30426394
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30426394 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426394 Supplier Toronto Research Chemicals Supplier No. TRCT30367510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1,2,5,6-tetramethylnaphthalene
CAS 2131-43-3
Molekylær formel C14 H16
Mængde 10 mg
InChI formel InChI=1S/C14H16/c1-9-5-7-14-12(4)10(2)6-8-13(14)11(9)3/h5-8H,1-4H3
SMIL Cc1ccc2c(C)c(C)ccc2c1C
IUPAC navn 1,2,5,6-tetramethylnaphthalene
Molekylvægt (g/mol) 184.277
Formel vægt 184.125
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.