Få mere at vide
1,2,4-Trimethylbenzene-d12, TRC
Artikelnummer. 30454676
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30454676 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454676 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79617710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,4-Trimethylbenzene D12
CAS 350818-61-0
Molekylær formel C9D12
Mængde 10 mg
Synonym Perdeuterated 1,2,4-trimethylbenzene
InChI formel InChI=1S/C9H12/c1-7-4-5-8(2)9(3)6-7/h4-6H,1-3H3/i1D3,2D3,3D3,4D,5D,6D
SMIL [2H]C([2H])([2H])C1=C([2H])C([2H])=C(C([2H])([2H])[2H])C([2H])=C1C([2H])([2H])[2H]
IUPAC navn 1,2,4-tris(methyl-d3)benzene-3,5,6-d3
Molekylvægt (g/mol) 132.27
Formel vægt 132.17