Få mere at vide
1,2,4-Trimethylbenzene-d12, CDN
Artikelnummer. 30317628
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30317628 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317628 Leverandør CDN Leverandørnr. CDND50621G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,4-Trimethylbenzene D12
CAS 350818-61-0
Molekylær formel C9D12
Mængde 1 g
Synonym Perdeuterated 1,2,4-trimethylbenzene
InChI formel InChI=1 S/C9H12/c1-7-4-5-8(2)9(3)6-7/h4-6 H,1-3H3/i1D3,2D3,3D3,4 D,5 D,6 D
SMIL [2 H]C([2 H])([2 H])C1=C([2 H])C([2 H])=C(C([2 H])([2 H])[2 H])C([2 H])=C1C([2 H])([2 H])[2 H]
IUPAC navn 1,2,4-tris(methyl-d3)benzene-3,5,6-d3
Molekylvægt (g/mol) 132.27
Formel vægt 132.17 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity