Få mere at vide
1,2,4-Trichlorobenzene-d3, TRC
Artikelnummer. 30450271
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30450271 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450271 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7742215MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,4-Trichlorobenzene-d3
CAS 2199-72-6
Molekylær formel C6 2H3 Cl3
Mængde 5 mg
Synonym Benzene-1,2,4-d3','3,5,6-trichloro-, Benzene-1,2,4-d3','trichloro- (7CI), 3,5,6-Trichlorobenzene-1,2,4-d3, 1,2,4-Trichlorobenzene-d3, 1,2,4-Trichlorobenzene D3
InChI formel InChI=1S/C6H3Cl3/c7-4-1-2-5(8)6(9)3-4/h1-3H/i1D,2D,3D
SMIL [2H]c1c([2H])c(Cl)c(Cl)c([2H])c1Cl
IUPAC navn 1,2,4-trichloro-3,5,6-trideuteriobenzene
Molekylvægt (g/mol) 184.47
Formel vægt 182.9489