Få mere at vide
1,2,4-Trichlorobenzene-d3, CDN
Artikelnummer. 30321260
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321260 0.25 g
30316205 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321260 Leverandør CDN Leverandørnr. CDND27060.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,4-Trichlorobenzene-d3
CAS 2199-72-6
Molekylær formel C6 2H3 Cl3
Mængde 0.25 g
Synonym Benzene-1,2,4-d3, 3,5,6-trichloro-, Benzene-1,2,4-d3, trichloro- (7 CI), 3,5,6-Trichlorobenzene-1,2,4-d3, 1,2,4-Trichlorobenzene-d3, 1,2,4-Trichlorobenzene D3
InChI formel InChI=1 S/C6H3Cl3/c7-4-1-2-5(8)6(9)3-4/h1-3 H/i1D,2 D,3 D
SMIL [2 H]c1c([2 H])c(Cl)c(Cl)c([2 H])c1Cl
IUPAC navn 1,2,4-trichloro-3,5,6-trideuteriobenzene
Molekylvægt (g/mol) 184.47
Formel vægt 182.9489 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity