Få mere at vide
1,2,4-Triazolo[4,3-a]pyridine-3(2H)-one, Mikromol™
Artikelnummer. 30049410
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049410 25 mg
17961856 100 mg
30461026 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30049410 Leverandør Mikromol Leverandørnr. MM0421.110025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,2,4-Triazolo[4,3-a]pyridine-3(2H)-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,4-Triazolo[4,3-a]pyridine-3(2H)-one
CAS 6969-71-7
Analyt- eller komponentnavne 1,2,4-Triazolo[4,3-a]pyridine-3(2H)-one
Forsendelsestilstand Room Temperature
Molekylær formel C6 H5 N3 O
Certificeringer/compliance ISO 17025
Synonym 1,2,4-Triazolo[4,3-a]pyridine-3(2H)-one
InChI formel InChI=1S/C6H5N3O/c10-6-8-7-5-3-1-2-4-9(5)6/h1-4H,(H,8,10)
SMIL O=C1NN=C2C=CC=CN12
IUPAC navn 2H-[1,2,4]triazolo[4,3-a]pyridin-3-one
Molekylvægt (g/mol) 135.12
Mængde 25 mg
Formel vægt 135.0433 g/mol