Få mere at vide
1,2,4-Triazolo[3,4-b]benzothiazole-5-methanol, TRC
Artikelnummer. 30456632
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456632 10 mg
30453989 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30456632 Leverandør Toronto Research Chemicals Leverandørnr. TRCT76737010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 69243-49-8
Molekylær formel C9H7N3OS
Molekylvægt (g/mol) 205.24
Synonym Tricyclazole-hydroxy, 1,2,4-Triazolo[3,4-b]benzothiazole-5-methanol (9CI, ACI)
IUPAC navn [1,2,4]triazolo[3,4-b][1,3]benzothiazol-8-ylmethanol
SMIL OCc1cccc2sc3nncn3c12
Tekniske data
Kemisk navn eller materiale 1,2,4-Triazolo[3,4-b]benzothiazole-5-methanol
Mængde 10 mg
InChI formel InChI=1S/C9H7N3OS/c13-4-6-2-1-3-7-8(6)12-5-10-11-9(12)14-7/h1-3,5,13H,4H2
Formel vægt 205.031