Få mere at vide
1,2,4-Triacetoxybenzene, 97%, Thermo Scientific™
Artikelnummer. 11491247
Skift visning
Mængde:
25 g
100 g
Pakningsstørrelse:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11491247 25 g 25g
11401257 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11491247 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B24208.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 613-03-6
Molekylær formel C12H12O6
Molekylvægt (g/mol) 252.222
MDL nummer MFCD00008700
InChI nøgle AESFGSJWSUZRGW-UHFFFAOYSA-N
Synonym 1,2,4-triacetoxybenzene, benzene-1,2,4-triyl triacetate, hydroxyhydroquinone triacetate, 1,2,4-benzenetriol, triacetate, pyrogallol a, 2-hydroxyhydroquinone triacetate, 1,2,4-benzenetriacetate, unii-903t7h7cp1, triacetate d'hydroxyhydroquinone, 2,4-bis acetyloxy phenyl acetate
PubChem CID 69169
IUPAC navn (3,4-diacetyloxyphenyl) acetate
SMIL CC(=O)OC1=CC(=C(C=C1)OC(=O)C)OC(=O)C
Tekniske data
Kemisk navn eller materiale 1,2,4-Triacetoxybenzene
Smeltepunkt 98°C to 100°C
Mængde 25 g
Beilstein 2138876
Formel vægt 252.22
Procent renhed 97%