Få mere at vide
1,2,4-Oxadiazol-3-amine, TRC
Artikelnummer. 30180429
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180429 10 mg
30179953 100 mg
30179790 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180429 Leverandør Toronto Research Chemicals Leverandørnr. TRCE59002810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 39512-64-6
Molekylær formel C2H3N3O
Molekylvægt (g/mol) 85.06
IUPAC navn 1,2,4-oxadiazol-3-amine
SMIL Nc1nocn1
Tekniske data
Kemisk navn eller materiale 1,2,4-Oxadiazol-3-amine
Mængde 10 mg
InChI formel InChI=1S/C2H3N3O/c3-2-4-1-6-5-2/h1H,(H2,3,5)
Formel vægt 85.0276