Få mere at vide
1-[2-(4-Morpholinyl)ethyl-d4]-3-(1-naphthoyl)indole JWH 200-d4, TRC
Artikelnummer. 30483315
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30483315 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483315 Leverandør Toronto Research Chemicals Leverandørnr. TRCM72376710MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[2-(4-Morpholinyl)ethyl-d4]-3-(1-naphthoyl)indole (JWH 200-d4)
CAS 1346600-73-4
Molekylær formel C25H20D4N2O2
Mængde 10 mg
Synonym [1-[2-(4-Morpholinyl)ethyl-d4]-1H-indol-3-yl]-1-naphthalenyl-methanone
InChI formel InChI=1S/C25H24N2O2/c28-25(22-10-5-7-19-6-1-2-8-20(19)22)23-18-27(24-11-4-3-9-21(23)24)13-12-26-14-16-29-17-15-26/h1-11,18H,12-17H2/i12D2,13D2
SMIL Cl.C[C@H](N)C(N)=O
Molekylvægt (g/mol) 388.49