Få mere at vide
1-[2-(4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinyl)ethyl] Ester Butanedioic Acid, TRC
Artikelnummer. 30012127
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012127 10 mg
30012752 100 mg
30011865 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012127 Leverandør Toronto Research Chemicals Leverandørnr. TRCH37143010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1872428-97-1
Molekylær formel C15 H27 N O5
Molekylvægt (g/mol) 301.379
IUPAC navn 4-[2-(4-hydroxy-2,2,6,6-tetramethylpiperidin-1-yl)ethoxy]-4-oxobutanoic acid
SMIL CC1(C)CC(O)CC(C)(C)N1CCOC(=O)CCC(=O)O
Tekniske data
Kemisk navn eller materiale 1-[2-(4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinyl)ethyl] Ester Butanedioic Acid
Mængde 10 mg
InChI formel InChI=1S/C15H27NO5/c1-14(2)9-11(17)10-15(3,4)16(14)7-8-21-13(20)6-5-12(18)19/h11,17H,5-10H2,1-4H3,(H,18,19)
Formel vægt 301.189