Få mere at vide
1,2,4-Benzenetricarboxylic Acid 1,2-Dihexyl Ester, TRC
Artikelnummer. 30333352
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30333352 250 mg
30329231 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30333352 Leverandør Toronto Research Chemicals Leverandørnr. TRCB190325250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,4-Benzenetricarboxylic Acid 1,2-Dihexyl Ester
CAS 142288-22-0
Molekylær formel C21H30O6
Mængde 250 mg
InChI formel InChI=1S/C21H30O6/c1-3-5-7-9-13-26-20(24)17-12-11-16(19(22)23)15-18(17)21(25)27-14-10-8-6-4-2/h11-12,15H,3-10,13-14H2,1-2H3,(H,22,23)
SMIL CCCCCCOC(=O)c1ccc(cc1C(=O)OCCCCCC)C(=O)O
IUPAC navn 3,4-bis(hexoxycarbonyl)benzoic acid
Molekylvægt (g/mol) 378.46
Formel vægt 378.2042