Få mere at vide
1-(2,4,6-Trimethoxybenzyl)piperazine Dihydrochloride, Mikromol™
Beskrivelse
1-(2,4,6-Trimethoxybenzyl)piperazine Dihydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-(2,4,6-Trimethoxybenzyl)piperazine Dihydrochloride |
| CAS | 113698-83-2 |
| Analyt- eller komponentnavne | 1-(2,4,6-Trimethoxybenzyl)piperazine Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H22 N2 O3 . 2 Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Trimetazidine Dihydrochloride Imp. F (EP), Trimetazidine Imp. F (EP), 1-(2,4,6-Trimethoxybenzyl)piperazine Dihydrochloride, Trimetazidine Dihydrochloride Imp. F (EP) as Dihydrochloride, Trimetazidine Dihydrochloride Impurity F as Dihydrochloride, Trimetazidine Impurity F as Dihydrochloride |
| InChI formel | InChI=1S/C14H22N2O3.2ClH/c1-17-11-8-13(18-2)12(14(9-11)19-3)10-16-6-4-15-5-7-16;;/h8-9,15H,4-7,10H2,1-3H3;2*1H |
| SMIL | Cl.Cl.COc1cc(OC)c(CN2CCNCC2)c(OC)c1 |
| IUPAC navn | 1-[(2,4,6-trimethoxyphenyl)methyl]piperazine;dihydrochloride |
| Vis mere |