Få mere at vide
1,2,4,5-Tetramethylbenzene-3,6-d2, CDN
Artikelnummer. 30319563
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319563 0.25 g
30314297 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319563 Leverandør CDN Leverandørnr. CDND22690.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,4,5-Tetramethylbenzene-3,6-D2
CAS 1859-01-4
Molekylær formel C10 D2 H12
Mængde 0.25 g
Synonym Benzene-1,4-d2, 2,3,5,6-tetramethyl- (8 CI, 9 CI), Benzene-p-d2, tetramethyl- (7 CI), 1,4-d2-Durol, 1,2,4,5-Tetramethylbenzene-3,6-d2, 1,2,4,5-Tetramethylbenzene-3,6-D2, Durene-d2 (ring-d2)
InChI formel InChI=1 S/C10H14/c1-7-5-9(3)10(4)6-8(7)2/h5-6 H,1-4H3/i5D,6 D
SMIL [2 H]c1c(C)c(C)c([2 H])c(C)c1C
IUPAC navn 1,4-dideuterio-2,3,5,6-tetramethylbenzene
Molekylvægt (g/mol) 136.2305
Formel vægt 136.1221 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity