Learn More
1,2,4,5-Benzenetetracarboxylic Acid, Sigma-Aldrich™
Product Code. 15888400
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15888400 250 g 250g
15878400 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15888400 Supplier Sigma Aldrich Supplier No. 8007840250
 Restricted Item
Forventet forsendelse: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89-05-4
Grade Research
Molecular Formula C6H2(COOH)4
Molecular Weight (g/mol) 254.15
Specifications
Melting Point 250°C to 260°C
Density 1.79 g/cm3 at 20°C
MDL Number MFCD00002471
Quantity 250 g
Synonym 1,2,4,5-Benzenetetracarboxylic acid, Benzene-1,2,4,5-tetracarboxylic acid, 1,2,4,5-Benzenetetracarboxylic acid