Få mere at vide
1,2,4,5-Benzenetetracarboxylic Acid-d6, CDN
Artikelnummer. 30309996
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309996 0.25 g
30316142 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309996 Leverandør CDN Leverandørnr. CDND24400.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,4,5-Benzenetetracarboxylic Acid-D6
CAS 344298-79-9
Molekylær formel C10 D6 O8
Mængde 0.25 g
Synonym 1,2,4,5-Benzene-3,6-d2-tetracarboxylic acid-d4 (9 CI), 1,2,4,5-Benzenetetracarboxylic acid-d6, 1,2,4,5-Benzenetetracarboxylic acid-D6, Pyromellitic acid-d6
InChI formel InChI=1 S/C10H6O8/c11-7(12)3-1-4(8(13)14)6(10(17)18)2-5(3)9(15)16/h1-2 H,(H,11,12)(H,13,14)(H,15,16)(H,17,18)/i1D,2 D/hD4
SMIL [2 H]OC(=O)c1c([2 H])c(C(=O)O[2 H])c(C(=O)O[2 H])c([2 H])c1C(=O)O[2 H]
IUPAC navn tetradeuterio 3,6-dideuteriobenzene-1,2,4,5-tetracarboxylate
Molekylvægt (g/mol) 260.1868
Formel vægt 260.0439 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity