missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,2,3-Trichloro-4-nitrobenzene, 97%

Catalog Number p-7023958
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
17700-09-3
C6H2Cl3NO2
226.437
MFCD00007063
BGKIECJVXXHLDP-UHFFFAOYSA-N
2,3,4-trichloronitrobenzene, trichloronitrobenzene, 2,3,4-trichloro-1-nitrobenzene, 4-nitro-1,2,3-trichlorobenzene, benzene, 1,2,3-trichloro-4-nitro, benzene, 4-nitro-1,2,3-trichloro, 1,2,3-trichloronitrobenzene, 1,2,3-tris chloranyl-4-nitro-benzene, pubchem8437, acmc-1bpdn
28697
1,2,3-trichloro-4-nitrobenzene
C1=CC(=C(C(=C1[N+](=O)[O-])Cl)Cl)Cl
This item is not returnable. View return policy
17700-09-3
C6H2Cl3NO2
226.437
MFCD00007063
BGKIECJVXXHLDP-UHFFFAOYSA-N
2,3,4-trichloronitrobenzene, trichloronitrobenzene, 2,3,4-trichloro-1-nitrobenzene, 4-nitro-1,2,3-trichlorobenzene, benzene, 1,2,3-trichloro-4-nitro, benzene, 4-nitro-1,2,3-trichloro, 1,2,3-trichloronitrobenzene, 1,2,3-tris chloranyl-4-nitro-benzene, pubchem8437, acmc-1bpdn
28697
1,2,3-trichloro-4-nitrobenzene
C1=CC(=C(C(=C1[N+](=O)[O-])Cl)Cl)Cl
Melting Point 53°C to 56°C
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN2811
Beilstein 2211951
Formula Weight 226.45
Percent Purity 97%
Chemical Name or Material 1,2,3-Trichloro-4-nitrobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'