missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1,2,3-Trichloro-4-nitrobenzene, 97%
Click to view available options
Quantity:
10 g
50 g
Specifications
Melting Point | 53°C to 56°C |
Flash Point | >110°C (230°F) |
Quantity | 50 g |
UN Number | UN2811 |
Beilstein | 2211951 |
Formula Weight | 226.45 |
Percent Purity | 97% |
Chemical Name or Material | 1,2,3-Trichloro-4-nitrobenzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'