missing translation for 'onlineSavingsMsg'
Learn More

1-[(2,3-Dihydro-1,4-benzodioxin-2-yl)carbonyl]piperazine Monohydrochloride, TRC

Product Code. 30373767
Change view
Click to view available options
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30373767 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373767 Supplier Toronto Research Chemicals Supplier No. TRCD44844525G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-[(2RS)-2,3-Dihydro-1,4-benzodioxin-2-ylcarbonyl]piperazine Hydrochloride
CAS 70918-74-0
Molekylær formel C13 H16 N2 O3 . Cl H
Mængde 25 g
Synonym 1-[(2RS)-2,3-Dihydro-1,4-benzodioxin-2-ylcarbonyl]piperazine Hydrochloride, Doxazosin Mesilate Imp. B (EP) as Hydrochloride, Methanone, (2,3-dihydro-1,4-benzodioxin-2-yl)-1-piperazinyl-, hydrochloride (1:1), Piperazine, 1-[(2,3-dihydro-1,4-benzodioxin-2-yl)carbonyl]-, monohydrochloride (9CI), 1,4-Benzodioxin, piperazine deriv., 1-[(2,3-Dihydro-1,4-benzodioxin-2-yl)carbonyl]piperazine Monohydrochloride, 1-(1,4-Benzodioxan-2-ylcarbonyl)piperazine hydrochloride
InChI formel InChI=1S/C13H16N2O3.ClH/c16-13(15-7-5-14-6-8-15)12-9-17-10-3-1-2-4-11(10)18-12;/h1-4,12,14H,5-9H2;1H
SMIL Cl.O=C(C1COc2ccccc2O1)N3CCNCC3
IUPAC navn 2,3-dihydro-1,4-benzodioxin-3-yl(piperazin-1-yl)methanone;hydrochloride
Molekylvægt (g/mol) 284.74
Formel vægt 284.0928
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.