missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3-Benzenetricarboxylic acid hydrate, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used in the synthesis and characterization and thermal behavior of hemimellitic acid complexes with lanthanides(III).
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used in the synthesis and characterization and thermal behavior of hemimellitic acid complexes with lanthanides(III).
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | ∼191°C (decomposition) |
Quantity | 5 g |
Beilstein | 2214816 |
Solubility Information | Slightly soluble in water. |
IUPAC Name | benzene-1,2,3-tricarboxylic acid hydrate |
Formula Weight | 210.14 (Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | 1,2,3-Benzenetricarboxylic acid hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'