Få mere at vide
1,2,3,9-Tetrahydro-9-(methyl-d3)-4H-carbazol-4-one, TRC
Artikelnummer. 30412905
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30412905 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412905 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29392225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,3,9-Tetrahydro-9-(methyl-d3)-4H-carbazol-4-one
CAS 1225443-54-8
Molekylær formel C13H10D3NO
Mængde 25 mg
InChI formel InChI=1S/C13H13NO/c1-14-10-6-3-2-5-9(10)13-11(14)7-4-8-12(13)15/h2-3,5-6H,4,7-8H2,1H3/i1D3
SMIL [2H]C([2H])([2H])n1c2CCCC(=O)c2c3ccccc13
IUPAC navn 9-(trideuteriomethyl)-2,3-dihydro-1H-carbazol-4-one
Molekylvægt (g/mol) 202.27
Formel vægt 202.1185