missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,6-Tetra-O-pivaloyl-Alpha-D-galactofuranoside, TRC

Product Code. 30455924
Change view
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30455924 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455924 Supplier Toronto Research Chemicals Supplier No. TRCT3063501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1,2,3,6-Tetra-O-pivaloyl-a-D-galactofuranoside
CAS 220017-49-2
Molekylær formel C26 H44 O10
Mængde 1 g
InChI formel InChI=1S/C26H44O10/c1-23(2,3)19(28)32-13-14-15(27)16(34-20(29)24(4,5)6)17(35-21(30)25(7,8)9)18(33-14)36-22(31)26(10,11)12/h14-18,27H,13H2,1-12H3/t14-,15+,16+,17-,18-/m1/s1
SMIL CC(C)(C)C(=O)OC[C@H]1O[C@H](OC(=O)C(C)(C)C)[C@H](OC(=O)C(C)(C)C)[C@@H](OC(=O)C(C)(C)C)[C@H]1O
IUPAC navn [(2R,3S,4S,5R,6R)-4,5,6-tris(2,2-dimethylpropanoyloxy)-3-hydroxyoxan-2-yl]methyl 2,2-dimethylpropanoate
Molekylvægt (g/mol) 516.62
Formel vægt 516.2934
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.