Få mere at vide
1,2,3,6,7,8-Hexachlorodibenzofuran, TRC
Artikelnummer. 30134537
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30134537 25 mg -
30005526 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134537 Leverandør Toronto Research Chemicals Leverandørnr. TRCH26510025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57117-44-9
Molekylær formel C12 H2 Cl6 O
Molekylvægt (g/mol) 374.86
Synonym Dibenzofuran, 1,2,3,6,7,8-hexachloro-, 1,2,3,6,7,8-Hexachlorodibenzofuran, 1,2,3,6,7,8-HxCDF, 123678-HxCDF, 2,3,4,7,8,9-Hexachlorodibenzofuran, F 121, PCDF 121, 1,2,3,6,7,8-HexaCDF, 123678-HexaCDF, 123678-Hexachlorodibenzofuran
IUPAC navn 1,2,3,6,7,8-hexachlorodibenzofuran
SMIL Clc1cc2oc3c(Cl)c(Cl)c(Cl)cc3c2c(Cl)c1Cl
Tekniske data
Kemisk navn eller materiale 1,2,3,6,7,8-Hexachlorodibenzofuran
Mængde 25 mg
InChI formel InChI=1S/C12H2Cl6O/c13-4-1-3-7-6(2-5(14)8(15)10(7)17)19-12(3)11(18)9(4)16/h1-2H
Formel vægt 371.8237