Få mere at vide
1,2,3,5-Tetrachlorobenzene-d2, TRC
Artikelnummer. 30138649
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138649 2 mg
30134929 1 mg
17907609 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30138649 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2919962MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2199-74-8
Molekylær formel C6 D2 Cl4
Molekylvægt (g/mol) 217.904
Synonym Benzene-m-d2, 2,4,5,6-tetrachloro- (8CI), Benzene-m-d2, tetrachloro- (7CI), 1,2,3,5-Tetrachlorobenzene-d2, 1,2,3,5-Tetrachlorobenzene-D2
IUPAC navn 1,2,3,5-tetrachloro-4,6-dideuteriobenzene
SMIL [2H]c1c(Cl)c([2H])c(Cl)c(Cl)c1Cl
Tekniske data
Kemisk navn eller materiale 1,2,3,5-Tetrachlorobenzene-d2
Mængde 2 mg
InChI formel InChI=1S/C6H2Cl4/c7-3-1-4(8)6(10)5(9)2-3/h1-2H/i1D,2D
Formel vægt 215.904