Få mere at vide
1,2,3,5-Tetrachlorobenzene-d2, CDN
Artikelnummer. 30321783
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321783 0.05 g -
30317436 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321783 Leverandør CDN Leverandørnr. CDND58700.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,3,5-Tetrachlorobenzene-d2
CAS 2199-74-8
Molekylær formel C6 D2 Cl4
Mængde 0.05 g
Synonym Benzene-m-d2, 2,4,5,6-tetrachloro- (8 CI), Benzene-m-d2, tetrachloro- (7 CI), 1,2,3,5-Tetrachlorobenzene-d2, 1,2,3,5-Tetrachlorobenzene-D2
InChI formel InChI=1 S/C6H2Cl4/c7-3-1-4(8)6(10)5(9)2-3/h1-2 H/i1D,2 D
SMIL [2 H]c1c(Cl)c([2 H])c(Cl)c(Cl)c1Cl
IUPAC navn 1,2,3,5-tetrachloro-4,6-dideuteriobenzene
Molekylvægt (g/mol) 217.904
Formel vægt 215.904 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity