missing translation for 'onlineSavingsMsg'
Learn More

1,2:3,5-Di-O-isopropylidene-alpha-D-xylofuranose, Thermo Scientific™

Catalog Number p-7043244
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
20881-04-3
C11H18O5
230.26
MFCD00063224
NKZDPBSWYPINNF-BZNPZCIMSA-N
1,2:3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-a-d-xylofuranose, alpha-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1s,2r,6r,8r-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 6.4.0.0 2 ,? dodecane, xylofuranose, 1,2:3,5-di-o-isopropylidene-, .alpha.-d, .alpha.-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1,2,3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1-o,2-o:3-o,5-o-diisopropylidene-alpha-d-xylofuranose, .alpha.-d-xylofuranose, 1,2,3,5-bis-o-1-methylethylidene
30337
(3aR,4aR,8aS,8bR)-2,2,7,7-tetramethyl-4a,5,8a,8b-tetrahydro-3aH-[1,3]dioxolo[3,4]furo[1,3-d][1,3]dioxine
CC1(OCC2C(O1)C3C(O2)OC(O3)(C)C)C
This item is not returnable. View return policy
20881-04-3
C11H18O5
230.26
MFCD00063224
NKZDPBSWYPINNF-BZNPZCIMSA-N
1,2:3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-d-xylofuranose, 1,2:3,5-di-o-isopropylidene-a-d-xylofuranose, alpha-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1s,2r,6r,8r-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 6.4.0.0 2 ,? dodecane, xylofuranose, 1,2:3,5-di-o-isopropylidene-, .alpha.-d, .alpha.-d-xylofuranose, 1,2:3,5-bis-o-1-methylethylidene, 1,2,3,5-di-o-isopropylidene-alpha-d-xylofuranose, 1-o,2-o:3-o,5-o-diisopropylidene-alpha-d-xylofuranose, .alpha.-d-xylofuranose, 1,2,3,5-bis-o-1-methylethylidene
30337
(3aR,4aR,8aS,8bR)-2,2,7,7-tetramethyl-4a,5,8a,8b-tetrahydro-3aH-[1,3]dioxolo[3,4]furo[1,3-d][1,3]dioxine
CC1(OCC2C(O1)C3C(O2)OC(O3)(C)C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Formula Weight 230.26
Physical Form Powder
Chemical Name or Material 1,2:3,5-Di-O-isopropylidene-alpha-D-xylofuranose
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'