missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetrahydro-1-naphthoic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.
Solubility
Soluble in methanol, and chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.
Solubility
Soluble in methanol, and chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Melting Point | 82°C to 85°C |
Quantity | 250 mg |
Solubility Information | Soluble in methanol,and chloroform. |
Formula Weight | 176.22 |
Percent Purity | 98% |
Chemical Name or Material | 1,2,3,4-Tetrahydro-1-naphthoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'