Få mere at vide
1,2,3,4-Tetrachlorobenzene-d2, TRC
Artikelnummer. 30416472
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30416472 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416472 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29095110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2,3,4-Tetrachlorobenzene-d2
CAS 2199-73-7
Molekylær formel C6 2H2 Cl4
Mængde 10 mg
Synonym Benzene-1,2-d2','3,4,5,6-tetrachloro- (9CI), Benzene-o-d2','3,4,5,6-tetrachloro- (8CI), Benzene-o-d2','tetrachloro- (7CI), 1,2,3,4-Tetrachlorobenzene D2
InChI formel InChI=1S/C6H2Cl4/c7-3-1-2-4(8)6(10)5(3)9/h1-2H/i1D,2D
SMIL [2H]c1c([2H])c(Cl)c(Cl)c(Cl)c1Cl
IUPAC navn 1,2,3,4-tetrachloro-5,6-dideuteriobenzene
Molekylvægt (g/mol) 217.9
Formel vægt 215.9036